Learn More
BURKLE INC Two-way valve, PVDF, Ø 0.35-0.43 in. (9-11 mm), NW 0.31 in. (8 mm), white
Catalog No. 5000025748
Encompass_Preferred
Two-way valves are used for transfer of liquids in order to release the flow, to block it or to change its direction.
Catalog No. 50-000-25748 Supplier BURKLE INC Supplier No. 86070100
Two-way valves are used for transfer of liquids in order to release the flow, to block it or to change its direction.