Learn More
BURKLE INC Two-way valve, PVDF, Ø 0.28-0.35 in. (7-9 mm), NW 0.24 in. (6 mm), white
Catalog No. 5000025946
Encompass_Preferred
Two-way valves are used for transfer of liquids in order to release the flow, to block it or to change its direction.
Catalog No. 50-000-25946 Supplier BURKLE INC Supplier No. 86070080
Two-way valves are used for transfer of liquids in order to release the flow, to block it or to change its direction.