Learn More
BURKLE INC Two-way valve, PP/PE, Ø 0.28-0.35 in. (7-9 mm), NW 0.24 in. (6 mm), red/white
Catalog No. 5000025598
Encompass_Preferred
Two-way valves are used for transfer of liquids in order to release the flow, to block it or to change its direction.
Catalog No. 50-000-25598 Supplier BURKLE INC Supplier No. 86050080
Two-way valves are used for transfer of liquids in order to release the flow, to block it or to change its direction.