Learn More
Andwin Scientific TWR DUAL PL PCH 3-1/2X8IN200PK
SDP
Catalog No. NC9508862
Encompass_Preferred
V. Mueller DualPeel Self-Sealing Pouch, Pressure-sensitive closure is prefolded and perforated to assure seal integrity. Blue-tinted viewing area. High porosity for fast sterilization while maintaining effective bacterial barrier.
Catalog No. NC9508862 Supplier Andwin Scientific Supplier No. 11213237
V. Mueller DualPeel Self-Sealing Pouch, Pressure-sensitive closure is prefolded and perforated to assure seal integrity. Blue-tinted viewing area. High porosity for fast sterilization while maintaining effective bacterial barrier.