Learn More
Apexbio Technology LLC TWS119 601514-19-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062460
Encompass_Preferred
TWS119 (CAS 601514-19-6) is a selective inhibitor of glycogen synthase kinase-3 (GSK-3 ) exhibiting an IC50 of approximately 30 nM Originally identified from a library of pyrrolopyrimidine-based compounds TWS119 binds GSK-3 with a Kd value around 126 nM By inhibiting GSK-3 activity TWS119 modulates downstream transcriptional events promoting neuronal differentiation in murine embryonic carcinoma (P19) and mouse embryonic stem cells through mechanisms distinct from classical Wnt signaling Due to this property TWS119 serves as a useful tool for research in neural differentiation and regenerative medicine
Catalog No. 50-000-62460 Supplier Apexbio Technology LLC Supplier No. B15405.1
TWS119 (CAS 601514-19-6) is a selective inhibitor of glycogen synthase kinase-3 (GSK-3 ) exhibiting an IC50 of approximately 30 nM Originally identified from a library of pyrrolopyrimidine-based compounds TWS119 binds GSK-3 with a Kd value around 126 nM By inhibiting GSK-3 activity TWS119 modulates downstream transcriptional events promoting neuronal differentiation in murine embryonic carcinoma (P19) and mouse embryonic stem cells through mechanisms distinct from classical Wnt signaling Due to this property TWS119 serves as a useful tool for research in neural differentiation and regenerative medicine