Learn More
Selleck Chemical LLC TWS119 10mM/1mL 601514-19-6
SDP
Catalog No. 501361408
Encompass_Preferred
TWS119 is a GSK-3? inhibitor with IC50 of 30 nM in a cell-free assay; capable of inducing neuronal differentiation and may be useful to stem cell biology. GSK-3? inhibition triggers autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1408 Supplier Selleck Chemical LLC Supplier No. S159010MM/1ML
TWS119 is a GSK-3? inhibitor with IC50 of 30 nM in a cell-free assay; capable of inducing neuronal differentiation and may be useful to stem cell biology. GSK-3? inhibition triggers autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.