Learn More
Abcam TWS119, GSK-3beta inhibitor, 2MG
Catalog No. 502706347
Encompass_Preferred
MW 318.3 Da, Purity >95%. Potent cell-permeable GSK-3β inhibitor (IC₅₀ = 30 nM). Induces stem cell neurogenesis in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6347 Supplier Abcam Supplier No. AB1420752MG
MW 318.3 Da, Purity >95%. Potent cell-permeable GSK-3β inhibitor (IC₅₀ = 30 nM). Induces stem cell neurogenesis in vitro.
The product is subject to the following: Abcam Restricted Use Statement