Learn More
Thermo Scientific™ TX-200 Swinging Bucket Rotor
Catalog No. 75003687 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Bucket
Bucket Lid
Replacement O-ring
Swinging Bucket Rotor
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
75-003-687 Replacement O-ring
75-003-658 Swinging Bucket Rotor
75-003-659 Bucket
75-003-660 Bucket Lid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 75-003-687 Supplier Thermo Scientific™ Supplier No. 75003687
  • $324.50
    $289.65 / Each of 1
    Save $34.85
    11% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Accommodate a wide range of processing volumes with the TX-200 4 x 180mL Swinging Bucket Rotor.
Accommodate a wide range of processing volumes with the Thermo Scientific™ TX-200 4 x 180mL Swinging Bucket Rotor.
Ordering Information:
Round buckets (75-003-659), biocontainment lids (75-003-660) and replacement O-rings (75-003-687) are sold separately. A variety of adapters are also available. Contact your sales representative for more information.
Compatible with:
Thermo Scientific Sorvall™ ST16, SL16, Sorvall Legend™ X1, Multifuge™ X1, Megafuge™ 16 Centrifuge Series
Recommended for:
Cell culture, blood separation, body fluids processing, microbiology, proteomics, genomics and molecular biology
TRUSTED_SUSTAINABILITY
Specifications
Type Replacement O-ring
For Use With (Equipment) Bucket Lids
Biocontainment Yes