Learn More
Medchemexpress LLC TX1-85-1 | 1603845-32-4 | 99.7% | 580.68 | 1 MG
SDP
Catalog No. 5000365209
Encompass_Preferred
TX1-85-1 is an irreversible Her3 (ErbB3) inhibitor with an IC50 of 23 nM. It acts as the first selective Her3 ligand, forming a covalent bond with Cys721 in the ATP-binding site of Her3. This compound induces partial degradation of Her3 protein and attenuates Her3-dependent signaling, showing anti-proliferation effects in various cell lines.
  • Irreversible Her3 (ErbB3) inhibitor with an IC50 of 23 nM
  • First selective Her3 ligand forming a covalent bond with Cys721
  • Induces partial degradation of Her3 protein
  • Attenuates Her3-dependent signaling
  • Demonstrates anti-proliferation EC50s of 9.9, 11.5, and 16.9 μM for Ovcar8, HCC827 GR6, and PC9 GR4 cells, respectively
Catalog No. 50-003-65209 Supplier Medchemexpress LLC Supplier No. 1MGHY100848
Add to Cart
Add to Cart
TX1-85-1 is an irreversible Her3 (ErbB3) inhibitor with an IC50 of 23 nM. It acts as the first selective Her3 ligand, forming a covalent bond with Cys721 in the ATP-binding site of Her3. This compound induces partial degradation of Her3 protein and attenuates Her3-dependent signaling, showing anti-proliferation effects in various cell lines.
  • Irreversible Her3 (ErbB3) inhibitor with an IC50 of 23 nM
  • First selective Her3 ligand forming a covalent bond with Cys721
  • Induces partial degradation of Her3 protein
  • Attenuates Her3-dependent signaling
  • Demonstrates anti-proliferation EC50s of 9.9, 11.5, and 16.9 μM for Ovcar8, HCC827 GR6, and PC9 GR4 cells, respectively