Learn More
Medchemexpress LLC HY-100848 5mg Medchemexpress, TX1-85-1 CAS:1603845-32-4 Purity:>98%
SDP
Catalog No. 501931969
Encompass_Preferred
Medchemexpress, HY-100848 5mg TX1-85-1 CAS:1603845-32-4 TX1-85-1 is an irreversible Her3 (ErbB3) inhibitor with an IC 50 of 23 nM and is also the first selective Her3 ligand , which forms a covalent bond with Cys721 located in the ATP-binding site of Her3. TX1-85-1 induces partial degradation of Her3 protein and attenuates Her3-dependent signaling [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1969 Supplier Medchemexpress LLC Supplier No. HY1008485MG
Medchemexpress, HY-100848 5mg TX1-85-1 CAS:1603845-32-4 TX1-85-1 is an irreversible Her3 (ErbB3) inhibitor with an IC 50 of 23 nM and is also the first selective Her3 ligand , which forms a covalent bond with Cys721 located in the ATP-binding site of Her3. TX1-85-1 induces partial degradation of Her3 protein and attenuates Her3-dependent signaling [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.