Learn More
Enzo Life Sciences TXB2 ELISA kit (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI901002
Encompass_Preferred
The TXB2 ELISA kit is a colorimetric competitive enzyme immunoassay kit with results in 2 in a convenient 96 well plate based assay. This kit is highly cited in peer-reviewed publications in a diverse assortment of sample types including mucosa, tissue, serum, plasma, urine, and much more. The TXB2 ELISA kit provides easy-to-use stable liquid color-coded reagents that reduce errors. Alternative Name: Thromboxane B2. Sensitivity: 10.54 pg/ml (range 13.7 - 10,000 pg/ml). Assay Time: <3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution
Catalog No. 50-658-198