Learn More
COAST Cutlery TXM50R All-In-One Light
Catalog No. 19030068
Change view
Click to view available options
Light Source:
LED
Catalog No. Light Source
19-030-068 LED
1 options
Catalog No. 19-030-068 Supplier COAST Cutlery Supplier No. 31188
Add to Cart
Add to Cart
Triple up on lighting functionality. Designed to maximize versatility, the rechargeable, hardhat-compatible TXM50R not only wears comfortably all shift as a headlamp but also transforms to become a pistol-grip handheld flashlight or a magnetic work light.
  • Built-In USB-C Port - With a built-in USB-C port, your battery can be charged using a USB-C cord. Some COAST models can be charged both inside or outside the light.
  • Dust Resistant - Take on any dirty work environment with this dust resistant light.
  • Hardhat Compatible - Rugged enough for the outside of the hardhat. COAST headlamps with Hardhat Compatible design include clips for attaching the light to the hat.
  • Magnetic Base - Attach the light to magnetic surfaces. Lights with a Magnetic Base have magnets located on their base that allow the light to be attached to magnetic surfaces for easy storage or hands-free use.
  • Pure Beam Focusing Optic - Pack two powerful beams into one and seamlessly transition - with no halos or hot spots - between short and long-range viewing. COAST's patented focusing technology adapts to your need. The Ultra View Flood beam delivers edge-to-edge clarity in a broad circle, and the BULLS-EYE™ Spot Beam sends a sharp ray of light at a distance with a transitional halo around the spot to increase the effective viewing area.
  • Turbo Mode - The XP lights not only features standard light modes but also COAST's signature Turbo Mode for overpowered light in critical situations.
  • Twist Focus - Dial in the shape of light you need for the situation. COAST's TWIST FOCUS™ system makes it easy to move between spot and flood beams by simply rotating the head of the flashlight.
Specifications
Light Source LED
Voltage 3.7 V
Color Black
Product Type Helmet-mounted Flashlight