Learn More
Aviva Systems Biology Corp TXN2 Antibody - middle region (ARP54910_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54910_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TXN2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TXN2. Alternate names: MT-TRX, MTRX, TRX2ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-61364