Learn More
Antibodies Online TXNDC12 Protein (AA 27-168) (His tag) 50ug
SDP
Catalog No. NC3360632
Encompass
Recombinant Human Thioredoxin Domain-Containing Protein 12/TXNDC12 is produced by our mammalian expression system in human cells. The target protein is expressed with sequence (His27-Leu168) of Human TXNDC12 fused with a polyhistidine tag at the C-terminus.
Catalog No. NC3360632 Supplier Antibodies Online Supplier No. ABIN1097634
Recombinant Human Thioredoxin Domain-Containing Protein 12/TXNDC12 is produced by our mammalian expression system in human cells. The target protein is expressed with sequence (His27-Leu168) of Human TXNDC12 fused with a polyhistidine tag at the C-terminus.