Learn More
Aviva Systems Biology Corp TXNDC13 Antibody - C-terminal region (ARP49630_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49630_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TXNDC13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TXNDC13. Alternate names: DJ971N18.2, KIAA1162, PDIA14, TXNDC13ApplicationIHC, WB. Species reactivity: Human, Pig
Catalog No. 50-014-60128