Learn More
Aviva Systems Biology Corp TXNDC4 Antibody - middle region (ARP58726_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58726_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TXNDC4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TXNDC4. Alternate names: ERP44, KIAA0573, PDIA10, TXNDC4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62403