Learn More
Aviva Systems Biology Corp TXNDC5 Antibody - N-terminal region (ARP58547_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58547_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TXNDC5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TXNDC5. Alternate names: ERP46, EndoPDI, MGC3178, UNQ364, HCC-2, STRF8, PDIA15, ENDOPDIApplicationWB. Species reactivity: Human
Catalog No. 50-014-62351