Learn More
Aviva Systems Biology Corp TXNIP Antibody - C-terminal region (ARP52193_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52193_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TXNIP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TXNIP. Alternate names: EST01027, HHCPA78, THIF, VDUP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-53684