Learn More
Aviva Systems Biology Corp TXNIP Antibody - middle region (ARP59054_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59054_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TXNIP. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TXNIP. Alternate names: EST01027, HHCPA78, THIF, VDUP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-568