Learn More
Medchemexpress LLC HY-115688 5mg Medchemexpress, TXNIP-IN-1 CAS:1268955-50-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1156885MG
Encompass_Preferred
Medchemexpress, HY-115688 5mg TXNIP-IN-1 CAS:1268955-50-5 TXNIP-IN-1 is TXNIP-TRX (thioredoxin-interacting protein- thioredoxin) complex inhibitor extracted from patent US20200085800A1, Compound 1. TXNIP-IN-1 can be used in the research of TXNIP-TRX complex associated metabolic disorder (diabetes), cardiovascular disease, or inflammatory disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9808