Learn More
Aviva Systems Biology Corp TXNRD3 Antibody - N-terminal region (ARP64748_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64748_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TXNRD3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human TXNRD3. Alternate names: TGR, TR2, TRXR3ApplicationWB. Species reactivity: Human, Mouse, Rabbit
Catalog No. 50-288-354