Learn More
Aviva Systems Biology Corp TXNRD3 Antibody - middle region (ARP64749_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64749_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TXNRD3. It was validated on Western Blot. . Alternate names: TGR, TR2, TRXR3ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-288-355