Learn More
Medchemexpress LLC HY-19736 5mg Medchemexpress, TY-52156 CAS:934369-14-9 Purity:>98%
SDP
Catalog No. 502030876
Encompass_Preferred
Medchemexpress, HY-19736 5mg TY-52156 CAS:934369-14-9 TY-52156 is a potent and selective S1P3 receptor antagonist with a Ki value of 110 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0876 Supplier Medchemexpress LLC Supplier No. HY197365MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-19736 5mg TY-52156 CAS:934369-14-9 TY-52156 is a potent and selective S1P3 receptor antagonist with a Ki value of 110 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.