Learn More
Turtle & Hughes Inc TY-RAP 1000 PER PKG
SDP
Catalog No. NC0005769
Encompass_Preferred
Cable Tie, Natural Polyamide (Nylon 6.6) for Temperatures up to 85 Degrees Celsius (185 F) for Indoor Applications, UL/EN/CSA62275 Type 2/21 Rated for AH-2 Plenum, Length of 284.4mm (11.08 Inches), Width of 3.71mm (0.14 Inches), Thickness of 1.31mm (0.05 Inches), Tensile Strength Rating of 135 Newtons (30 pounds), Bulk Pack
Catalog No. NC0005769 Supplier Turtle & Hughes Inc Supplier No. TY26M
Cable Tie, Natural Polyamide (Nylon 6.6) for Temperatures up to 85 Degrees Celsius (185 F) for Indoor Applications, UL/EN/CSA62275 Type 2/21 Rated for AH-2 Plenum, Length of 284.4mm (11.08 Inches), Width of 3.71mm (0.14 Inches), Thickness of 1.31mm (0.05 Inches), Tensile Strength Rating of 135 Newtons (30 pounds), Bulk Pack