Learn More
Andwin Scientific TY-RAP HITMP 4IN 1000/PK
SDP
Catalog No. NC9067137
Encompass_Preferred
The Thomas & Betts - TYHT23M - High Temperature Light Green Nylon Cable Tie features a length of 3.62 and a tensile of 18lb. This product provides unlimited tensioning range for a perfect fit. It is constructed of extra high temperature nylon that allows these ties to operate up to 302 F while still quicker to install and less expensive compared to metallic fasteners, lacing cord or tape. This is sold in a pack of 1000.
Catalog No. NC9067137 Supplier Andwin Scientific Supplier No. TYHT23M1000
The Thomas & Betts - TYHT23M - High Temperature Light Green Nylon Cable Tie features a length of 3.62 and a tensile of 18lb. This product provides unlimited tensioning range for a perfect fit. It is constructed of extra high temperature nylon that allows these ties to operate up to 302 F while still quicker to install and less expensive compared to metallic fasteners, lacing cord or tape. This is sold in a pack of 1000.