Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuflow Systems Versilon Tubing, FEP, 3/8 in ID, 9/16 in OD

Catalog No. NC3584527
Encompass_Preferred

Versilon SE-200 is an FEP-lined, clear flexible tubing designed for inert chemical transfer. It combines a smooth FEP inner liner with a flexible outer polymer to provide chemical resistance, visual clarity, and a tight bend radius for routing in process and laboratory systems.

  • FEP inner liner for chemical inertness and low extractables.
  • Clear construction for visual fluid inspection.
  • 3/8 in inner diameter, 9/16 in outer diameter, 3/32 in wall thickness.
  • Minimum bend radius 3.5 in for tight routing.
  • Maximum operating temperature 165 °F (74 °C).
  • Maximum working pressure 50 psi at 73 °F; 25 psi at 160 °F.
  • Durometer hardness 66 Shore A; tensile strength 2,200 psi; ultimate elongation 390%.
  • Common applications: chemical processing, semiconductor processing, photographic and adhesive transfer.

Catalog No. NC3584527 Supplier Accuflow Systems Supplier No. AJD00028
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Versilon SE-200 is an FEP-lined, clear flexible tubing designed for inert chemical transfer. It combines a smooth FEP inner liner with a flexible outer polymer to provide chemical resistance, visual clarity, and a tight bend radius for routing in process and laboratory systems.

  • FEP inner liner for chemical inertness and low extractables.
  • Clear construction for visual fluid inspection.
  • 3/8 in inner diameter, 9/16 in outer diameter, 3/32 in wall thickness.
  • Minimum bend radius 3.5 in for tight routing.
  • Maximum operating temperature 165 °F (74 °C).
  • Maximum working pressure 50 psi at 73 °F; 25 psi at 160 °F.
  • Durometer hardness 66 Shore A; tensile strength 2,200 psi; ultimate elongation 390%.
  • Common applications: chemical processing, semiconductor processing, photographic and adhesive transfer.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.