Learn More
MSC AbilityOne Floor Pad, Polyester, Buffing, Red, 19 in Diameter
Catalog No. 502839659
Encompass
These red polyester floor pads are designed for use with rotary disk floor maintenance machines operating up to 750 rpm. They are reusable, corrosion-resistant, and conform to Commercial Item Description A-A-3004, Type III. Each pad measures 19 inch in diameter and features a 3-3/8 inch center hole.
  • Effectively removes light scuffs and dirt.
  • Revitalizes the appearance of floors.
  • Safe for most types of floors.
  • Cleans when wet, buffs when dry.
  • Conforms to the floor's contours, ensuring thorough cleaning and buffing.
  • Used to buff very soft finishes or polish soft waxes on wood floor.
  • Made in the U.S.A.
  • Compliant to the U.S. Trade Agreement Act (TAA).
Catalog No. 50-283-9659 Supplier MSC Supplier No. 89463780
These red polyester floor pads are designed for use with rotary disk floor maintenance machines operating up to 750 rpm. They are reusable, corrosion-resistant, and conform to Commercial Item Description A-A-3004, Type III. Each pad measures 19 inch in diameter and features a 3-3/8 inch center hole.
  • Effectively removes light scuffs and dirt.
  • Revitalizes the appearance of floors.
  • Safe for most types of floors.
  • Cleans when wet, buffs when dry.
  • Conforms to the floor's contours, ensuring thorough cleaning and buffing.
  • Used to buff very soft finishes or polish soft waxes on wood floor.
  • Made in the U.S.A.
  • Compliant to the U.S. Trade Agreement Act (TAA).