Learn More
MSC AbilityOne Floor Pad, Nylon, Buffing, 19 in, Tan
Catalog No. 502839743
Encompass
These 19 in diameter nylon buffing floor pads in tan are designed for concentrated weight, rotary disk floor machines operating between 150-350 rpm. They feature a 3-3/8 in center hole and measure 11/16 in thick. These washable, reusable, and corrosion-resistant pads are suitable for all resilient tile, terrazzo, marble, and ceramic floors, conforming to Commercial Item Description A-A-3004, Type I. Each box contains 5 pads.
  • Effectively removes light scuffs and dirt.
  • Revitalizes the appearance of floors.
  • Safe for most types of floors.
  • Cleans when wet, buffs when dry.
  • Conforms to the floor's contours, ensuring thorough cleaning and buffing.
  • Washable and reusable.
  • Corrosion-resistant.
  • Designed for rotary disk floor machines rated between 150-350 rpm.
  • Conforms to Commercial Item Description A-A-3004, Type I.
Catalog No. 50-283-9743 Supplier MSC Supplier No. 89463772
These 19 in diameter nylon buffing floor pads in tan are designed for concentrated weight, rotary disk floor machines operating between 150-350 rpm. They feature a 3-3/8 in center hole and measure 11/16 in thick. These washable, reusable, and corrosion-resistant pads are suitable for all resilient tile, terrazzo, marble, and ceramic floors, conforming to Commercial Item Description A-A-3004, Type I. Each box contains 5 pads.
  • Effectively removes light scuffs and dirt.
  • Revitalizes the appearance of floors.
  • Safe for most types of floors.
  • Cleans when wet, buffs when dry.
  • Conforms to the floor's contours, ensuring thorough cleaning and buffing.
  • Washable and reusable.
  • Corrosion-resistant.
  • Designed for rotary disk floor machines rated between 150-350 rpm.
  • Conforms to Commercial Item Description A-A-3004, Type I.