Learn More
MSC AbilityOne Floor Pad, Polyester, Buffing, Red, 16 in Diameter
Catalog No. 502839590
Encompass
This floor pad is designed for use with concentrated weight, rotary disk floor maintenance machines operating up to 750 rpm. It is reusable and corrosion-resistant, conforming to Commercial Item Description A-A-3004, Type III, and features a 3-3/8 in center hole.
  • Effectively removes light scuffs and dirt
  • Revitalizes the appearance of floors
  • Safe for most types of floors
  • Cleans when wet, buffs when dry
  • Conforms to the floor's contours, ensuring thorough cleaning and buffing
Catalog No. 50-283-9590 Supplier MSC Supplier No. 89462543
This floor pad is designed for use with concentrated weight, rotary disk floor maintenance machines operating up to 750 rpm. It is reusable and corrosion-resistant, conforming to Commercial Item Description A-A-3004, Type III, and features a 3-3/8 in center hole.
  • Effectively removes light scuffs and dirt
  • Revitalizes the appearance of floors
  • Safe for most types of floors
  • Cleans when wet, buffs when dry
  • Conforms to the floor's contours, ensuring thorough cleaning and buffing