Learn More
MSC SKILCRAFT Floor Pad, White Polishing 4100 Series, 21 in, Polyester
Catalog No. 502837128
Encompass
This floor pad, part of the White Polishing 4100 Series, is designed for buffing very soft finishes or polishing soft waxes on wood floors. Crafted from 3M floor pad materials, this 21 in diameter, Type III polyester polishing pad is optimized for machines operating between 175 and 600 rpm.
  • Use to buff very soft finishes.
  • Ideal for polishing soft waxes on wood floors.
  • Constructed with 3M floor pad materials.
  • Designed for 175-600 rpm machine operation.
  • Features a 21 in diameter.
  • Comes in boxes of 5 pads.
  • White polishing pad for light cleaning.
  • Made from polyester material.
Catalog No. 50-283-7128 Supplier MSC Supplier No. 89463095
This floor pad, part of the White Polishing 4100 Series, is designed for buffing very soft finishes or polishing soft waxes on wood floors. Crafted from 3M floor pad materials, this 21 in diameter, Type III polyester polishing pad is optimized for machines operating between 175 and 600 rpm.
  • Use to buff very soft finishes.
  • Ideal for polishing soft waxes on wood floors.
  • Constructed with 3M floor pad materials.
  • Designed for 175-600 rpm machine operation.
  • Features a 21 in diameter.
  • Comes in boxes of 5 pads.
  • White polishing pad for light cleaning.
  • Made from polyester material.