Learn More
MSC AbilityOne SKILCRAFT Floor Pad, Polyester, Polishing, White, 14 inch Diameter
Catalog No. 502839745
Encompass
These floor pads are designed for use with concentrated weight, rotary disk floor maintenance machines operating up to 750 rpm. They are reusable, corrosion-resistant, and conform to Commercial Item Description A-A-3004, Type III.
  • Pads are reusable and corrosion-resistant.
  • Conforms to Commercial Item Description A-A-3004, Type III.
  • Suitable for rotary disk floor maintenance machines up to 750 rpm.
  • Features a 3-3/8 inch center hole.
  • Includes 5 pads per box.
Catalog No. 50-283-9745 Supplier MSC Supplier No. 89463277
These floor pads are designed for use with concentrated weight, rotary disk floor maintenance machines operating up to 750 rpm. They are reusable, corrosion-resistant, and conform to Commercial Item Description A-A-3004, Type III.
  • Pads are reusable and corrosion-resistant.
  • Conforms to Commercial Item Description A-A-3004, Type III.
  • Suitable for rotary disk floor maintenance machines up to 750 rpm.
  • Features a 3-3/8 inch center hole.
  • Includes 5 pads per box.