Learn More
MSC SKILCRAFT Floor Pad, Polyester, Stripping, 19 inch Diameter, Brown
Catalog No. 502837171
Encompass
These polyester floor pads are designed for use with concentrated weight, rotary disk floor maintenance machines up to 750 rpm. They are highly effective for aggressively removing old layers of floor finish, wax, and sealers, and are also suitable for deep scrubbing. These durable pads are reusable, corrosion-resistant, and conformable for uneven surfaces, allowing for efficient cleaning with both wet and dry methods.
  • Aggressively removes old layers of floor finish, wax, and sealers
  • Effective for deep scrubbing
  • Reusable and corrosion-resistant
  • Conformable for uneven surfaces
  • Works with wet or dry methods
  • Meets Commercial Item Description A-A-3004, Type III
  • Features a 3-3/8 inch center hole
  • Packaged with 5 pads per box
Catalog No. 50-283-7171 Supplier MSC Supplier No. 89461792
These polyester floor pads are designed for use with concentrated weight, rotary disk floor maintenance machines up to 750 rpm. They are highly effective for aggressively removing old layers of floor finish, wax, and sealers, and are also suitable for deep scrubbing. These durable pads are reusable, corrosion-resistant, and conformable for uneven surfaces, allowing for efficient cleaning with both wet and dry methods.
  • Aggressively removes old layers of floor finish, wax, and sealers
  • Effective for deep scrubbing
  • Reusable and corrosion-resistant
  • Conformable for uneven surfaces
  • Works with wet or dry methods
  • Meets Commercial Item Description A-A-3004, Type III
  • Features a 3-3/8 inch center hole
  • Packaged with 5 pads per box