Learn More
DuPont™ Tychem™ 10000 Hooded Coverall
Catalog No. 18888840 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
5X-Large
6X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-888-840 5X-Large
18-888-841 6X-Large
18-888-842 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-888-840 Supplier DuPont™ Supplier No. TK128TLY5X000200
Add to Cart
Add to Cart
This item is not returnable. View return policy
Hooded Coverall offers a high level of protection against toxic, corrosive gases and liquid chemicals
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Manufactured under specifications that do not contain natural rubber latex
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
  • Attached respirator-fit hood with elastic around face opening, designed to cover neck and chin and fit around respirator face mask
  • Strong zipper for tough, flexible closure
  • Longer zipper extends to chin for complete coverage of neck area
  • Storm flap covers zipper which can be sealed by the wearer with two adhesive strips to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
TRUSTED_SUSTAINABILITY
Specifications
Color Lime Yellow
Material Tychem
Size 5X-Large
Ankle Style Boot Overflaps
Closure Type Zipper
Cuff Style Elastic
Disposable Single-use
Gender Unisex
Latex Free Yes
Neck Style Hooded
No. of Pockets None
Quantity 2 Pk.
Resistance Liquid
Seam Type Taped
Sterility Non-sterile