Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ 10000 Encapsulated Level A Suit with MSA Dual Purpose

Catalog No. 18888876 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-888-876 X-Large
18-888-859 2X-Large
18-888-869 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-888-876 Supplier DuPont™ Supplier No. TK555TLYXL00017M
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Suit with MSA dual purpose offers a high level of protection against toxic, corrosive gases and liquid chemicals

  • 10000 Encapsulated Level A Suit with MSA Dual Purpose garment design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment
  • Integrated socks composed of garment material
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Two exhaust valves release higher pressure inside encapsulating garments while simultaneously helping to prevent inward leakage of external vapors or particles into the garment
  • Internal adjustable waist belt system for support and improved fit
  • Additional barrier material sewn at knees and over taped for added abrasion resistance when kneeling or crawling
  • Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052
  • Manufactured under specifications that do not contain natural rubber latex
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
  • Rear entry design keeps closure away from direct contact with frontal chemical exposure and provides ability to change air bottle on SCBA without removing the entire garment
  • Expanded back to accommodate self-contained air breathing apparatus (SCBA)
  • Vapor tight zipper
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Attached internal gloves composed of five layer laminate film for a broad range of chemical protection
  • Butyl gloves provide mechanical and physical durability to help protect inner glove
  • Gloves are attached with vapor tight ring and clamp system and are replaceable
TRUSTED_SUSTAINABILITY

Specifications

Color Lime Yellow
For Use With (Application) Industrial, Chemical Handling, Emergency Response
Material Tychem
Size X-Large
Ankle Style Boot Overflaps
Closure Type Zipper
Product Type Encapsulated Level A Suit
Seam Type Taped
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.