Learn More
DuPont™ Tychem™ 10000 Encapsulated Level A Suit with 892 Outer Gloves
Shop All DuPont Personal Protection ProductsDescription
- Encapsulated Level A garment design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment
- Integrated socks composed of garment material
- Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
- Two exhaust valves release higher pressure inside encapsulating garments while simultaneously helping to prevent inward leakage of external vapors or particles into the garment
- One-way valves are positive pressure and open on demand
- Valves include splash covers made of barrier material to help prevent liquid intrusion
- Located on left back of head and right lower back
- Internal adjustable waist belt system for support and improved fit
- Additional barrier material sewn at knees and over taped for added abrasion resistance when kneeling or crawling
- Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052.
- Manufactured under specifications that do not contain natural rubber latex
- Pass-thrus are optional and can be installed upon request when ordering. Some suits are available with optional pass-thrus to accommodate the following SCBA manufacturers: Scott, Interspiro, MSA, Draeger, Survivair™
- Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
- Double taped seams provide strong chemical resistance against heavy liquid splashes
- Sewn seam is covered, both inside and outside of garment with compatible chemical-resistant material by heat-sealing.
- Expanded anti-fog face shield provides wide 220 degree viewing angle
- EX face shield is three-layer laminate composed of 40 mil PVC, 5 mil TeflonTM, 20 mil PVC
- Rear entry design keeps closure away from direct contact with frontal chemical exposure and provides ability to change air bottle on SCBA without removing the entire garment
- Expanded back to accommodate self-contained air breathing apparatus (SCBA)
- Vapor tight zipper. Extra long to increase garment opening and aid donning and doffing
- Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
- Provides at least 30 minutes of protection against over 320 chemical challenges
- Chosen by hazmat responders worldwide
- Available in high-visibility lime yellow
- Garments include fully encapsulated, vapor protective Level A suit and liquid-splash protective Level B suits as well as coveralls
- Attached internal gloves composed of five layer laminate film for a broad range of chemical protection
- Butyl gloves provide mechanical and physical durability to help protect inner glove
- Gloves are attached with vapor tight ring and clamp system and are replaceable
Specifications
Specifications
| Color | Lime Yellow |
| For Use With (Application) | Industrial, Chemical Handling, Emergency Response |
| Material | Tychem |
| Size | X-Large |
| Ankle Style | Boot Overflaps |
| Closure Type | Zipper |
| Product Type | Encapsulated Level A Suit |
| Seam Type | Taped |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.