Learn More
DuPont™ Tychem™ 10000 Training Suit
Catalog No. 18888878 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
3X-Large
4X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-888-878 3X-Large
18-888-879 4X-Large
18-888-880 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-888-878 Supplier DuPont™ Supplier No. TK586SLY3X000100
Add to Cart
Add to Cart
This item is not returnable. View return policy
10000 training suits offers a high level of protection against toxic, corrosive gases and liquid chemicals
  • Tychem™ 10000 fully encapsulated training suits offer an economical and effective option for HazMat training. Same fabric and garment design as standard Tychem™ 10000 suits
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Extra wide (EX) single layer faceshield (20 mil PVC) for wide viewing angle
  • Front entry design allows wearer to partially don garment (maintain standby position) without assistance
  • Integrated butyl gloves
  • Internal adjustable waist belt system for support and improved fit
  • Clearly labeled for training use only
  • Manufactured under specifications that do not contain natural rubber latex
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
TRUSTED_SUSTAINABILITY
Specifications
Color Lime Yellow
Material Tychem
Size 3X-Large
Ankle Style Boot Overflaps
Closure Type Zipper
Seam Type Serged