Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ 10000 Training Suit

Catalog No. 18888889 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
4X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-888-889 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-888-889 Supplier DuPont™ Supplier No. TK587SLY4X000100
  • $543.00
    $421.65 / Case of 1

    Save $121.35

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Offers a high level of protection against toxic, corrosive gases and liquid chemicals

  • Tychem™10000 fully encapsulated training suits offer an economical and effective option for hazmat training.
  • Same fabric and garment design as standard Tychem™ 10000 suits
  • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Extra wide (EX) single layer face shield (20 mil PVC) for wide viewing angle
  • Rear entry design keeps closure away from direct contact with frontal chemical exposure and provides ability to change air bottle on SCBA without removing the entire garment
  • Integrated butyl gloves
  • Internal adjustable waist belt system for support and improved fit
  • Clearly labeled for training use only
  • Manufactured under specifications that do not contain natural rubber latex
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
  • D13491807
TRUSTED_SUSTAINABILITY

Specifications

Color Lime Yellow
For Use With (Application) Hazmat and Emergency Response Training
Material Tychem
Size 4X-Large
Ankle Style Boot Overflaps
Closure Type Zipper
Product Type Encapsulated Training Suit
Seam Type Serged
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.