Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ 10000 Training Suit

Catalog No. 18888888 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-888-888 X-Large
18-888-881 2X-Large
18-888-882 3X-Large
18-888-883 4X-Large
18-888-884 5X-Large
18-888-885 6X-Large
18-888-886 Large
18-888-887 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 18-888-888 Supplier DuPont™ Supplier No. TK586TLYXL000100
  • $709.00
    $549.65 / Case of 1

    Save $159.35

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Offers a high level of protection against toxic, corrosive gases and liquid chemicals

  • Tychem™ 10000 fully encapsulated training suits offer an economical and effective option for hazmat training
  • Taped seams provide strong chemical resistance against heavy liquid splashes
    • Sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
  • Extra wide (EX) single layer face shield (20 mil PVC) for wide viewing angle
  • Front entry design allows wearer to partially don garment (maintain standby position) without assistance
  • Integrated butyl gloves
  • Internal adjustable waist belt system for support and improved fit
  • Clearly labeled for training use only
  • Manufactured under specifications that do not contain natural rubber latex
  • Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
TRUSTED_SUSTAINABILITY

Specifications

Color Lime Yellow
For Use With (Application) Hazmat and Emergency Response Training
Material Tychem
Size X-Large
Ankle Style Boot Overflaps
Closure Type Zipper
Product Type Encapsulated Training Suit
Seam Type Taped
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.