Learn More
DuPont™ Tychem™ 10000FR Encapsulated Level A Suit with Hansen Fitting
Catalog No. 18888757 Shop All DuPont Personal Protection Products
This item is not returnable. View return policy
Catalog No. 18-888-757 Supplier DuPont™ Supplier No. RF600TSV2X00017S
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Tychem™ 10000 FR is a top choice for chemical protection in emergency situations.
Certified to NFPA 1991 in a single-skin construction, Tychem® 10000 FR garments are composed of a patented, heavy-duty aluminized fabric made of a DuPont™ Nomex™ and DuPont™ Kevlar™ blend, laminated to a multi-layer barrier film. Tychem® 10000 FR fabrics are tested by a third party laboratory to assess its chemical permeation resistance.
  • Provides at least 30 minutes of protection against over 285 chemical challenges
  • Ideally suited for industrial and HAZMAT situations
  • Tychem™ 10000 FR is high-visibility silver
  • A limited-use “single skin” garment offers broad chemical holdout
Certifications
NFPA 1991 with Flash Fire and Liquified Gas
TRUSTED_SUSTAINABILITY
Specifications
Material Tychem