Learn More
DuPont™ Tychem™ 10000FR Encapsulated Level A Suit with MSA Dual Purpose
Catalog No. 18888768 Shop All DuPont Personal Protection Products
This item is not returnable. View return policy
Catalog No. 18-888-768
Supplier: DuPont™ RF600TSVXL00017M
This item is not returnable. View return policy
Tychem™ 10000 FR is a top choice for chemical protection in emergency situations.
Certified to NFPA 1991 in a single-skin construction, Tychem® 10000 FR garments are composed of a patented, heavy-duty aluminized fabric made of a DuPont™ Nomex™ and DuPont™ Kevlar™ blend, laminated to a multi-layer barrier film. Tychem® 10000 FR fabrics are tested by a third party laboratory to assess its chemical permeation resistance.
  • Provides at least 30 minutes of protection against over 285 chemical challenges
  • Ideally suited for industrial and HAZMAT situations
  • Tychem™ 10000 FR is high-visibility silver
  • A limited-use “single skin” garment offers broad chemical holdout
Certifications
NFPA 1991 with Flash Fire and Liquified Gas
TRUSTED_SUSTAINABILITY
Specifications
Material Tychem
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.