Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ 2000 Coverall (all)

Catalog No. 19162762 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Seam Type:
Bound
Serged
Taped
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Seam Type
19-162-762 X-Large Taped
19-162-759 5X-Large Taped
18-178C Large Serged
18-178F 3X-Large Serged
19-162-757 3X-Large Taped
19-162-760 Large Taped
18-178D X-Large Serged
19-162-758 4X-Large Taped
19-053921 2X-Large Bound
18-178E 2X-Large Serged
19-017-977B Medium Bound
19-120-2582 5X-Large Serged
18-178B Medium Serged
19-162-756 2X-Large Taped
19-017976 X-Large Bound
19-067-017 4X-Large Serged
19-162-761 Medium Taped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
This item is not returnable. View return policy
Catalog No. 19-162-762 Supplier DuPont™ Supplier No. QC125TYLXL000400
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Flexible and durable light liquid splash protection

  • Tychem™ QC is polyethylene-coated Tyvek™ fabric fabric provides at least 30 minutes of protection against 42 chemical challenges
    • Laydown collar
    • Elastic opening for tighter fit at wrist
    • Elastic opening for tighter fit at ankle
    • Yellow for high visibility
    • 10 mil thickness
  • QC 125B
    • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
    • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • QC 125S
    • Multiple interlocking threads are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • QC 125T
    • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing
    • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper

Compliance

Meets ASTM™ D3776, ASTM D1777, ASTM 1117, ASTM D3786, ASTM D5034 standards.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemical Protective Clothing
Size X-Large
Gender Unisex
Material Tychem
Color Yellow
Disposable Single-use
Waist Style Non-elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Standard Collar
Closure Type Front Zipper
Seam Type Taped
Sterility Non-sterile
Resistance Chemicals, Non-toxic Particulates
For Use With (Application) Petroleum refining, pulp and paper manufacturing, food processing, chemical processing, and pharmaceutical manufacturing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.