Description
- Knee-length apron/gown design with attached sleeves.
- Manufactured under specifications that do not contain natural rubber latex
- Provides enhanced frontal protection.
- 44 in. long for neck-to-knee coverage.
- Open back for wearer comfort
- Hook and loop closure.
- Ties at waist.
- Elastic opening for tighter fit at wrist.
- Available in multiple sizes.
- Taped seams provide strong chemical resistance against heavy liquid splashes
Specifications
Specifications
| Type | Chemical-resistant Apron |
| Size | Small |
| Material | Tychem |
| Color | Green |
| Disposable | Single-use |
| Closure Type | Hook and Loop, Waist Ties |
| Sterility | Non-sterile |
| Seam Type | Taped |