Learn More
DuPont™ Tychem™ 2000 SFR Hooded Coverall
Catalog No. 19147775 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-147-775 Small
19-147-776 Medium
19-147-777 Large
19-147-778 X-Large
19-147-779 2X-Large
19-147-780 3X-Large
19-147-781 4X-Large
19-147-782 5X-Large
19-147-783 6X-Large
19-147-784 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 19-147-775 Supplier DuPont™ Supplier No. QS127TGRSM000400
Add to Cart
Add to Cart
This item is not returnable. View return policy
Provides chemical and secondary flame protection in a lightweight garment.
  • Respirator-fit hood lined with ProShield™ 6 SFR fabric.
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
  • Covered, braided elastic at hood.
  • Nylon zipper with large metal pull
  • Chin flap with double-sided adhesive tape
  • Storm flap covers zipper which can be sealed by the wearer with double-sided adhesive strip to prevent intrusion at zipper.
  • Covered, braided elastic openings at wrist and ankle for tighter fit.
  • Taped seams.
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Manufactured under specifications that do not contain natural rubber latex
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hooded Coveralls with Socks
Size Small
Gender Unisex
Material Tychem
Color Green
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Hood Type Respirator Fit (Fabric-lined)
Closure Type Zipper
Seam Type Taped
Resistance Chemicals