Learn More
DuPont™ Tychem™ 4000 Coverall (all)
Catalog No. 17500149 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Ankle Style:
Elastic
Catalog No. Ankle Style
17-500-149 Elastic
1 options
This item is not returnable. View return policy
Catalog No. 17-500-149 Supplier DuPont™ Supplier No. SL125BWH6X001200
Add to Cart
Add to Cart
This item is not returnable. View return policy
DuPont™ Tychem™ 4000 Coveralls provide at least 30 minutes of protection against >124 chemical challenges in a range of chemical environments
  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Laydown collar
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
TRUSTED_SUSTAINABILITY
Specifications
Product Type Chemical Protective Clothing
Size 6X-Large
Gender Unisex
Material Tychem
Color White
Disposable Single-use
Waist Style Non-elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Standard Collar
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Resistance Chemicals, Bloodborne Pathogens
No. of Pockets None