Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ 4000 Hooded Coverall

Catalog No. 19120694B Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
7X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-120-694B Medium
18-643 Large
18-642 X-Large
18-641 2X-Large
19-120-694C 3X-Large
18-999-592 4X-Large
17-500-151 7X-Large
17-500-150 6X-Large
18-999-593 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-120-694B Supplier DuPont™ Supplier No. SL127BWHMD001200
Add to Cart
Add to Cart
This item is not returnable. View return policy

DuPont™ Tychem™ 4000 Coveralls provide at least 30 minutes of protection against >124 chemical challenges in a range of chemical environments.

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Attached hood with elastic around face opening.
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex

Compliance

Meets ASTM™ D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786; 16 CFR 1610 Standard for Flammability of Clothing Textiles; MIL-STD-282 requirements.

TRUSTED_SUSTAINABILITY

Specifications

Size Medium
Gender Unisex
Material Tychem
Color White
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper
Seam Type Bound
Certifications/Compliance ASTM D3776, ASTM D5034, ASTM D1777, ASTM F739, ASTM D3786, 16 CFR 1610, MIL-STD-282
Size (English) Chest 35.25 to 38.75 in.
Size (English) Inseam 27.5 in.
Size (Metric) Chest 89.53 to 98.42 cm
Thickness 12 mil
For Use With (Application) Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
Length (English) Sleeve 33.75 in.
Length (Metric) Sleeve 85.72 cm
Width (English) Chest 24.25 in.
Height (English) 5.3 to 5.7 ft.
Height (Metric) 1.6 to 1.73 m
Weight (English) 0.22 lb.
Weight (Metric) 0.1 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.