Learn More
MSC Dupont Tychem 4000 Coverall, Chemical and Bloodborne Pathogen Protection, 2X-Large, White, 6/Case
Catalog No. 5000687179
Encompass
The DuPont™ Tychem® 4000 coverall is designed for protection against chemical and bloodborne pathogen hazards. Constructed with Tychem® 4000 fabric, which utilizes Saranex 23-P film laminated to Tyvek® fabric, this garment offers a durable barrier that resists abrasion. It is suitable for applications including hazardous remediation, chemical mixing, emergency medical response, paint spraying, and nuclear environments.
  • Provides at least 30 minutes of protection against more than 124 chemical challenges
  • Features taped seams and a storm flap with adhesive closure for enhanced containment
  • Includes a standard fit hood, elastic wrists, and attached socks
  • Designed with a pin lock slider zipper pull
  • Constructed from 3.6 oz./yd2 material with a thickness of 12 mils
  • Excludes components containing natural rubber latex
Catalog No. 50-006-87179 Supplier MSC Supplier No. 07793474
Add to Cart
Add to Cart
The DuPont™ Tychem® 4000 coverall is designed for protection against chemical and bloodborne pathogen hazards. Constructed with Tychem® 4000 fabric, which utilizes Saranex 23-P film laminated to Tyvek® fabric, this garment offers a durable barrier that resists abrasion. It is suitable for applications including hazardous remediation, chemical mixing, emergency medical response, paint spraying, and nuclear environments.
  • Provides at least 30 minutes of protection against more than 124 chemical challenges
  • Features taped seams and a storm flap with adhesive closure for enhanced containment
  • Includes a standard fit hood, elastic wrists, and attached socks
  • Designed with a pin lock slider zipper pull
  • Constructed from 3.6 oz./yd2 material with a thickness of 12 mils
  • Excludes components containing natural rubber latex