Learn More
DuPont™ Tychem™ 5000 Bib Overall
Catalog No. 17500107 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
4X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-500-107 4X-Large
17-500-108 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-500-107 Supplier DuPont™ Supplier No. C3360TTN4X000600
Add to Cart
Add to Cart
This item is not returnable. View return policy
Bib Overall with Open Ankles provides protection against a broad range of chemicals and situations involving liquid splash.
  • Bib style overall with adjustable webbing straps
  • Typically worn in combination with C3670T TN jacket or in appplications were potential exposure is below waist
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks
  • NFPA 1992 certified
TRUSTED_SUSTAINABILITY
Specifications
Closure Type Adjustable Webbing Straps with Snap-lock
Color Tan
Material Tychem
Seam Type Taped
Size 4X-Large
Suspender Adjustable Webbing Straps with Buckle Closure
Ankle Style Open