Learn More
DuPont™ Tychem™ 5000 Coverall (all)
Catalog No. 17500055 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Medium
2X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-500-055 2X-Large
17-500-056 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-500-055 Supplier DuPont™ Supplier No. C3184TTN2X000600
Add to Cart
Add to Cart
This item is not returnable. View return policy
DuPont™ Tychem™ 5000 Coverall with Collar, Attached Dual Layer Gloves provides protection against a broad range of chemicals and situations involving liquid splash.
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
  • Tychem™ CPF3 HD garments were designed with extensive input from first responders to ensure that HAZMAT, law enforcement, military and hospital personnel get the protection they need from chemical warfare agents (including Sarin, Mustard, and Lewisite) and toxic industrial chemicals (TIC's).
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Mandarin collar designed to fit tightly around hooded PAPR
  • Larger zipper pull is easier to grasp for easier donning and doffing
  • Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
  • Attached internal gloves composed of five layer laminate film for a broad range of chemical protection
  • Neoprene outer gloves deliver high tactility and dexterity to handle tools or weapons
  • Integrated socks composed of garment material
TRUSTED_SUSTAINABILITY
Specifications
Size 2X-Large
Gender Unisex
Material Tychem
Color Tan
Ankle Style Attached Socks, Boot Overflaps
Closure Type Zipper
Seam Type Taped
Includes Attached Dual Layer Gloves
Style Collared