Learn More
DuPont™ Tychem™ 5000 Coverall (all)
Catalog No. 17500039 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
Small
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-500-039 Small
17-500-038 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-500-039 Supplier DuPont™ Supplier No. C3125TTNSM000600
Add to Cart
Add to Cart
This item is not returnable. View return policy
DuPont™ Tychem™ 5000 Coverall with Collar, Elastic Wrists and Ankles provides protection against a broad range of chemicals and situations involving liquid splash.
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Laydown collar
  • Strong zipper for tough, flexible closure
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Chemical Protective Clothing
Size Small
Gender Unisex
Material Tychem
Color Tan
Disposable Single-use
Waist Style Non-elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Standard
Closure Type Storm Flap with Adhesive Closure
Seam Type Taped
Sterility Non-sterile
Resistance Chemicals, Non-toxic Particulates
Style Collared