Learn More
DuPont™ Tychem™ 5000 Hooded Coverall
Catalog No. 17500036 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Size:
6X-Large
7X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-500-036 6X-Large
17-500-037 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-500-036 Supplier DuPont™ Supplier No. C3122TTN6X000600
Add to Cart
Add to Cart
This item is not returnable. View return policy
DuPont™ TYCHEM™ 5000 Coverall with Standard Fit Hood, Attached Socks provides protection against a broad range of chemicals and situations involving liquid splash.
  • Zipper features a semi-automatic locking slider allowing the puller to stay down and in a locked position
  • Taped seams provide strong chemical resistance against heavy liquid splashes. A sewn seam is covered with a strip of compatible chemical-resistant material through heat sealing.
  • Attached hood with elastic around face opening.
  • Strong zipper for tough, flexible closure
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Integrated socks composed of garment material
  • Manufactured under specifications that do not contain natural rubber latex
  • Meets the U.S. industry requirements for blood (ASTM F1670) and viral penetration (ASTM F1671) protecting against several bloodborne pathogen exposure risks.
TRUSTED_SUSTAINABILITY
Specifications
Size 6X-Large
Gender Unisex
Material Tychem
Color Tan
Ankle Style Attached Socks
Cuff Style Elastic
Closure Type Zipper
Seam Type Taped